Merck
c-KIT oncogene expression in PRKAR1A-mutant adrenal cortex.
Endocrine-related cancer (2020-08-02)
Kiran Nadella, Fabio R Faucz, Constantine A Stratakis, Kiran Nadella, Fabio R Faucz, Constantine A Stratakis
ABSTRAKT
Protein kinase A (PKA) regulatory subunit type 1A (PRKAR1A) defects lead to primary pigmented nodular adrenocortical disease (PPNAD). The KIT protooncogene (c-KIT) is not known to be expressed in the normal adrenal cortex (AC). In this study, we investigated the expression of c-KIT and its ligand, stem cell factor (SCF), in PPNAD and other cortisol-producing tumors of the adrenal cortex. mRNA and protein expression, by qRT-PCR, immunohistochemistry (IHC) and immunoblotting (IB), respectively, were studied. We then tested c-KIT and SCF responses to PRKAR1A introduction and PKA stimulation in adrenocortical cell lines CAR47 and H295R, which were also treated with the KIT inhibitor, imatinib mesylate (IM). Mice xenografted with H295R cells were treated with IM. There was increased c-KIT mRNA expression in PPNAD; IHC showed KIT and SCF immunoreactivity within certain nodular areas in PPNAD. IB data was consistent with IHC and mRNA data. PRKAR1A-deficient CAR47 cells expressed c-KIT; this was enhanced by forskolin and lowered by PRKAR1A reintroduction. Knockdown of PKA's catalytic subunit (PRKACA) by siRNA reduced c-KIT levels. Treatment of the CAR47 cells with IM resulted in reduced cell viability, growth arrest, and apoptosis. Treatment with IM of mice xenografted with H295 cells inhibited further tumor growth. We conclude that c-KIT is expressed in PPNAD, an expression that appears to be dependent on PRKAR1A and/or PKA activity. In a human adrenocortical cell line and its xenografts in mice, c-KIT inhibition decreased growth, suggesting that c-KIT inhibitors may be a reasonable alternative therapy to be tested in PPNAD, when other treatments are not optimal.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Imatinib