Merck
Tm7sf2 gene promotes adipocyte differentiation of mouse embryonic fibroblasts and improves insulin sensitivity.
Biochimica et biophysica acta. Molecular cell research (2020-10-31)
Leonardo Gatticchi, Maya Petricciuolo, Paolo Scarpelli, Lara Macchioni, Lanfranco Corazzi, Rita Roberti
ABSTRAKT
Adipogenesis is a finely orchestrated program involving a transcriptional cascade coordinated by CEBP and PPAR family members and by hormonally induced signaling pathways. Alterations in any of these factors result into impaired formation of fully differentiated adipocytes. Tm7sf2 gene encodes for a Δ(14)-sterol reductase primarily involved in cholesterol biosynthesis. Furthermore, TM7SF2 modulates the expression of the master gene of adipogenesis PPARγ, suggesting a role in the regulation of adipose tissue homeostasis. We investigated the differentiation of Tm7sf2-/- MEFs into adipocytes, compared to Tm7sf2+/+ MEFs. Tm7sf2 expression was increased at late stage of differentiation in wild type cells, while Tm7sf2-/- MEFs exhibited a reduced capacity to differentiate into mature adipocytes. Indeed, Tm7sf2-/- MEFs had lower neutral lipid accumulation and reduced expression of adipogenic regulators. At early stage, the reduction in C/EBPβ expression impaired mitotic clonal expansion, which is needed by preadipocytes for adipogenesis induction. At late stage, the expression and activity of C/EBPα and PPARγ were inhibited in Tm7sf2-/- cells, leading to the reduced expression of adipocyte genes like Srebp-1c, Fasn, Scd-1, Adipoq, Fabp4, and Glut4. Loss of the acquisition of adipocyte phenotype was accompanied by a reduction in the levels of Irs1, and phosphorylated Akt and ERK1/2, indicating a blunted insulin signaling in differentiating Tm7sf2-/- cells. Moreover, throughout the differentiation process, increased expression of the antiadipogenic Mmp3 was observed in MEFs lacking Tm7sf2. These findings indicate Tm7sf2 as a novel factor influencing adipocyte differentiation that could be relevant to adipose tissue development and maintenance of metabolic health.