Merck
Tau phosphorylation and cleavage in ethanol-induced neurodegeneration in the developing mouse brain.
Neurochemical research (2010-01-06)
Mariko Saito, Goutam Chakraborty, Rui-Fen Mao, Sun-Mee Paik, Csaba Vadasz, Mitsuo Saito
ABSTRAKT
Previous studies indicated that ethanol-induced neurodegeneration in postnatal day 7 (P7) mice, widely used as a model for the fetal alcohol spectrum disorders, was accompanied by glycogen synthase kinase-3beta (GSK-3beta) and caspase-3 activation. Presently, we examined whether tau, a microtubule associated protein, is modified by GSK-3beta and caspase-3 in ethanol-treated P7 mouse forebrains. We found that ethanol increased phosphorylated tau recognized by the paired helical filament (PHF)-1 antibody and by the antibody against tau phosphorylated at Ser199. Ethanol also generated tau fragments recognized by an antibody against caspase-cleaved tau (C-tau). C-tau was localized in neurons bearing activated caspase-3 and fragmented nuclei. Over time, cell debris and degenerated projections containing C-tau appeared to be engulfed by activated microglia. A caspase-3 inhibitor partially blocked C-tau formation. Lithium, a GSK-3beta inhibitor, blocked ethanol-induced caspase-3 activation, phosphorylated tau elevation, C-tau formation, and microglial activation. These results indicate that tau is phosphorylated by GSK-3beta and cleaved by caspase-3 during ethanol-induced neurodegeneration in the developing brain.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Tau Antibody, a.a. 210-241, clone Tau-5, ascites fluid, clone Tau-5, Chemicon®
Sigma-Aldrich
Przeciwciało anty-Tau, rozszczepione przez kaspazę (skrócone przy Asp421), clone tau-C3, Chemicon®, from mouse