Merck
The role of somatostatin and its receptors (sstr2, sstr5) in the contractility of gilt inflamed uterus.
Research in veterinary science (2020-10-02)
Barbara Jana, Jarosław Całka, Marta Czajkowska
ABSTRAKT
We studied the inflammation effect on somatostatin receptors subtypes 2 (sstr2) and 5 (sstr5) expression in myometrium and somatostatin influence alone or with sstr2 and sstr5 antagonists on the contractility of gilt inflamed uterus. On day 3 of the estrous cycle, either E.coli suspension (E.coli group) or saline (SAL group) were injected into uterine horns. In the control pigs (CON group), only laparotomy was performed. Eight days later, in the E.coli group developed severe acute endometritis. In this group, myometrial sstr2 mRNA expression lowered and protein expression increased compared to other groups. Compared to period before somatostatin administration, somatostatin did not change tension in myometrium and endometrium/myometrium of three groups, reduced amplitude and frequency in the CON and SAL groups, and increased amplitude and decreased frequency in the E.coli group. In this group, amplitude was increased by somatostatin compared to other groups. In the CON and SAL groups, sstr2 eliminated inhibitory somatostatin effect on amplitude, while sstr5 antagonist reversed inhibitory somatostatin effect on amplitude. In the E.coli group, sstr2 antagonist reversed stimulatory somatostatin effect on amplitude, while in sstr5 antagonist presence stimulatory somatostatin effect was more deepened compared to somatostatin action alone. After using sstr2 antagonist more deepened inhibitory somatostatin effect on frequency in the CON and E.coli groups was found. Sstr5 antagonist partly eliminated inhibitory somatostatin effect on frequency in the SAL group. Summarizing, the uterine inflammation increases the myometrial sstr2 protein expression; somatostatin raises amplitude of the inflamed uterus acting by sstr2, while drops this parameter by sstr5.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triton X-100, laboratory grade