Merck
Pairing a prognostic target with potential therapeutic strategy for head and neck cancer.
Oral oncology (2020-10-23)
Sze Min Lek, Ke Li, Qiu Xuan Tan, Nicholas B Shannon, Wai Har Ng, Josephine Hendrikson, Joey W S Tan, Hui Jun Lim, Yudong Chen, Kelvin K N Koh, Thakshayeni Skanthakumar, Xue Lin Kwang, Fui Teen Chong, Hui Sun Leong, Gerald Tay, Natascha Ekawati Putri, Tony Kiat Hon Lim, Jacqueline S G Hwang, Mei Kim Ang, Daniel S W Tan, Ngian Chye Tan, Hiang Khoon Tan, Oi Lian Kon, Khee Chee Soo, N Gopalakrishna Iyer, Chin-Ann J Ong
ABSTRAKT
We have previously identified and validated a panel of molecular prognostic markers (ATP13A3, SSR3, and ANO1) for Head and Neck Squamous Cell Carcinoma (HNSCC). The aim of this study was to investigate the consequence of ATP13A3 dysregulation on signaling pathways, to aid in formulating a therapeutic strategy targeting ATP13A3-overexpressing HNSCC. Gene Set Enrichment Analysis (GSEA) was performed on HNSCC microarray expression data (Internal local dataset [n = 92], TCGA [n = 232], EMBL [n = 81]) to identify pathways associated with high expression of ATP13A3. Validation was performed using immunohistochemistry (IHC) on tissue microarrays (TMAs) of head and neck cancers (n = 333), staining for ATP13A3 and phosphorylated Aurora kinase A (phospho-T288). Short interfering RNA was used to knockdown ATP13A3 expression in patient derived HNSCC cell lines. Protein expression of ATP13A3 and Aurora kinase A was then assessed by immunoblotting. GSEA identified Aurora kinase pathway to be associated with high expression of ATP13A3 (p = 0.026). The Aurora kinase pathway was also associated with a trend towards poor prognosis and tumor aggressiveness (p = 0.086, 0.094, respectively). Furthermore, the immunohistochemical staining results revealed a significant association between Aurora kinase activity and high ATP13A3 expression (p < 0.001). Knockdown of ATP13A3 in human head and neck cell lines showed decrease in Aurora kinase A levels. Tumors with high ATP13A3 are associated with high Aurora kinase activity. This suggests a potential therapeutic role of Aurora kinase inhibitors in a subset of poor prognosis HNSCC patients with overexpression of ATP13A3.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human ATP13A3