Merck
Effects of dietary gamma-cyclodextrin on voluntary activity and muscle strength in mice.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society (2020-09-30)
S Wupper, A Fischer, K Luersen, I R Ipharraguerre, K Chikamoto, T Furune, Y Ishida, K Terao, G Rimbach
ABSTRAKT
Gamma-cyclodextrin (γCD) is a cyclic oligosaccharide consisting of eight α-(1,4)-linked glucopyranose subunits, which is often used in the food and pharmaceutical industries. However, little is known regarding the metabolic activity of "empty" γCD per se. Therefore, in the present study young C57BL/6 male mice received a control diet (CON) or an experimental diet that was supplemented with 12.88% γCD exchanged against corn starch. After 6 weeks of treatment, the voluntary wheel running activity was monitored and the muscle strength of mice was measured by employing Kondziela's inverted screen test and forelimb grip strength assay. The γCD-treated mice covered a significantly larger distance per night (CON 8.6 km, γCD 12.4 km) and were significantly longer active (CON 340 min, γCD 437 min). Moreover, γCD-treated mice significantly performed better at the inverted screen test indicated by an enhanced Kondziela score (CON 3.10, γCD 4.63). These data suggest that dietary γCD leads to an increased endurance. We also found a slightly anti-glycemic effect of γCD during oral glucose tolerance test. However, our mice from the γCD group exhibited no difference in terms of GLUT2 protein level in ileum tissue nor increased muscle glycogen storage. Furthermore, γCD exhibited no DPP-4 inhibitory activity in vitro. By analysing candidate muscle genes and proteins related to endurance and muscle performance we did not observe any differences in terms of Sirt1, Pgc1α, Cpt1b, Mef2c, Myh1 and Myh2 gene expression levels as well as total oxidative phosphorylation (OXPHOS), mtTFA and GLUT4 protein expression levels in skeletal muscle in response to γCD. We could not fully establish the exact underlying molecular mechanisms of the fitness improvement by dietary γCD which warrants further investigations.