Merck
  • Zinc Oxide nanoparticles induce oxidative and proteotoxic stress in ovarian cancer cells and trigger apoptosis Independent of p53-mutation status.
Zinc Oxide nanoparticles induce oxidative and proteotoxic stress in ovarian cancer cells and trigger apoptosis Independent of p53-mutation status.
Applied surface science (2020-02-12)
Achuth Padmanabhan, M Kaushik, R Niranjan, JoAnne S Richards, Brandon Ebright, G Devanand Venkatasubbu
ABSTRAKT
Ovarian cancer continues to be the most lethal among gynecological malignancies and the major cause for cancer-associated mortality among women. Limitations of current ovarian cancer therapeutics is highlighted by the high frequency of drug-resistant recurrent tumors and the extremely poor 5-year survival rates. Zinc oxide nanoparticles (ZnO-NPs) have shown promise in various biomedical applications including utility as anti-cancer agents. Here, we describe the synthesis and characterization of physical properties of ZnO-NPs of increasing particle size (15 nm - 55 nm) and evaluate their benefits as an ovarian cancer therapeutic using established human ovarian cancer cell lines. Our results demonstrate that the ZnO-NPs induce acute oxidative and proteotoxic stress in ovarian cancer cells leading to their death via apoptosis. The cytotoxic effect of the ZnO-NPs was found to increase slightly with a decrease in nanoparticle size. While ZnO-NPs caused depletion of both wild-type and gain-of-function (GOF) mutant p53 protein in ovarian cancer cells, their ability to induce apoptosis was found to be independent of the p53-mutation status in these cells. Taken together, these results highlight the potential of ZnO-NPs to serve as an anti-cancer therapeutic agent for treating ovarian cancers independent of the p53 mutants of the cancer cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution