Merck
Binary Fate Choice between Closely Related Interneuronal Types Is Determined by a Fezf1-Dependent Postmitotic Transcriptional Switch.
Neuron (2019-12-10)
Yi-Rong Peng, Rebecca E James, Wenjun Yan, Jeremy N Kay, Alex L Kolodkin, Joshua R Sanes
ABSTRAKT
Many neuronal types occur as pairs that are similar in most respects but differ in a key feature. In some pairs of retinal neurons, called paramorphic, one member responds to increases and the other to decreases in luminance (ON and OFF responses). Here, we focused on one such pair, starburst amacrine cells (SACs), to explore how closely related neuronal types diversify. We find that ON and OFF SACs are transcriptionally distinct prior to their segregation, dendritic outgrowth, and synapse formation. The transcriptional repressor Fezf1 is selectively expressed by postmitotic ON SACs and promotes the ON fate and gene expression program while repressing the OFF fate and program. The atypical Rho GTPase Rnd3 is selectively expressed by OFF SACs and regulates their migration but is repressed by Fezf1 in ON SACs, enabling differential positioning of the two types. These results define a transcriptional program that controls diversification of a paramorphic pair.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Vesicular Acetylcholine Transporter (VAChT) Antibody, from goat, purified by affinity chromatography
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-Green Fluorescent Protein Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat