Merck
  • Gallic acid affects intestinal-epithelial-cell integrity and selected amino-acid uptake in porcine in vitro and ex vivo permeability models.
Gallic acid affects intestinal-epithelial-cell integrity and selected amino-acid uptake in porcine in vitro and ex vivo permeability models.
The British journal of nutrition (2020-11-05)
Marco Tretola, Giuseppe Bee, Paolo Silacci
ABSTRAKT
Gallic acid (GA) is widely used as a dietary supplement due to several health-promoting effects, although its effects on intestinal-epithelial-cell integrity and transport remain mostly unknown. The present study aims to clarify the effects of GA on tight junctions and intestinal nutrient uptake through in vitro and ex vivo models. Both intestinal porcine enterocyte cell line-J2 cells and porcine middle-jejunum segments were treated with 5 (T5), 25 (T25) and 50 (T50) µm GA and mounted in Ussing chambers to determine transepithelial resistance (TEER), claudin-1 (CLDN1), occludin (OCLN), zonula occludens-1 (ZO-1) protein (in tissues and cells) and mRNA (in cells) expression. In addition, uptake of l-glutamate (l-Glut), l-arginine (l-Arg), l-lysine (l-Lys) and l-methionine (l-Meth) together with cationic-amino-acid transporter-1 (CAT-1) and excitatory-amino-acid transporter-3 (EAAT3) expression was evaluated. No apoptosis was observed in GA-treated cells, but TEER and CLDN1 protein abundance was lower with T50 compared with untreated cells. l-Arg and l-Lys uptake was greater with T5 than with T25 and T50. Ex vivo, T50 decreased the TEER values and the protein levels of CLDN1, OCLN and ZO-1, whereas T5 and T25 only decreased CLDN1 protein expression compared with untreated tissues. Moreover, T25 increased l-Glut and l-Arg uptake, the latter confirmed by an increased protein expression of CAT-1. GA influences intestinal uptake of the tested cationic amino acids at low concentrations and decreases the intestinal-cell barrier function at high concentrations. Similarities were observed between in vitro and ex vivo, but different treatment times and structures must be considered.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Koniugat peroksydazy koziej przeciw mysiej IgG, lyophilized, Calbiochem®