Merck
Connexin 36 Mediates Orofacial Pain Hypersensitivity Through GluK2 and TRPA1.
Neuroscience bulletin (2020-10-18)
Qian Li, Tian-Le Ma, You-Qi Qiu, Wen-Qiang Cui, Teng Chen, Wen-Wen Zhang, Jing Wang, Qi-Liang Mao-Ying, Wen-Li Mi, Yan-Qing Wang, Yu-Xia Chu, Qian Li, Tian-Le Ma, You-Qi Qiu, Wen-Qiang Cui, Teng Chen, Wen-Wen Zhang, Jing Wang, Qi-Liang Mao-Ying, Wen-Li Mi, Yan-Qing Wang, Yu-Xia Chu
ABSTRAKT
Trigeminal neuralgia is a debilitating condition, and the pain easily spreads to other parts of the face. Here, we established a mouse model of partial transection of the infraorbital nerve (pT-ION) and found that the Connexin 36 (Cx36) inhibitor mefloquine caused greater alleviation of pT-ION-induced cold allodynia compared to the reduction of mechanical allodynia. Mefloquine reversed the pT-ION-induced upregulation of Cx36, glutamate receptor ionotropic kainate 2 (GluK2), transient receptor potential ankyrin 1 (TRPA1), and phosphorylated extracellular signal regulated kinase (p-ERK) in the trigeminal ganglion. Cold allodynia but not mechanical allodynia induced by pT-ION or by virus-mediated overexpression of Cx36 in the trigeminal ganglion was reversed by the GluK2 antagonist NS102, and knocking down Cx36 expression in Nav1.8-expressing nociceptors by injecting virus into the orofacial skin area of Nav1.8-Cre mice attenuated cold allodynia but not mechanical allodynia. In conclusion, we show that Cx36 contributes greatly to the development of orofacial pain hypersensitivity through GluK2, TRPA1, and p-ERK signaling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
2,2,2-Tribromoethanol, 97%
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 antibody produced in mouse, clone NE14, ascites fluid
Sigma-Aldrich
Anti-Cre Recombinase Antibody, clone 2D8, ascites fluid, clone 2D8, Chemicon®
Sigma-Aldrich
Mefloquine hydrochloride, ≥98% (HPLC), powder