Merck
Innovative Hyperbranched Polybenzoxazine-Based Graphene Oxide-Poly(amidoamines) Nanomaterials.
Polymers (2020-10-25)
Elena Iuliana Bîru, Sorina Alexandra Gârea, Horia Iovu
ABSTRAKT
The covalent functionalization of graphene oxide (GO) surface with hyperbranched benzoxazine (BZ) structures has been achieved using poly(amidoamine) dendrimers (PAMAM) of different generations. By increasing the PAMAM generation, multiple benzoxazine rings were synthesized decorating the GO layers. The polymerization process and the exfoliation behavior were investigated. The novel BZ-functionalized GO hybrid materials were characterized by a combination of techniques such as FT-IR, XPS, and 1H-NMR for the confirmation of benzoxazine formation onto the GO layer surfaces. Raman and XRD investigation showed that the GO stacking layers are highly disintegrated upon functionalization with hyperbranched benzoxazine monomers, the exfoliation being more probably to occur when lower PAMAM generation (G) is involved for the synthesis of hybrid GO-BZ nanocomposites. The polymerization of BZ rings may occur either between the BZ units from the same dendrimer molecule or between BZ units from different dendrimer molecules, thus influencing the intercalation/exfoliation of GO. DSC data showed that the polymerization temperature strongly depends on the PAMAM generation and a significant decrease of this value occurred for PAMAM of higher generation, the polymerization temperature being reduced with ~10 °C in case of GO-PAMAM(G2)-BZ. Moreover, the nanoindentation measurements showed significant mechanical properties improvement in case of GO-PAMAM(G2)-BZ comparing to GO-PAMAM(G0)-BZ in terms of Young modulus (from 0.536 GPa to 1.418 GPa) and stiffness (from 3617 N/m to 9621 N/m).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)