Merck
  • Endoplasmic reticulum stress controls iron metabolism through TMPRSS6 repression and hepcidin mRNA stabilization by RNA-binding protein HuR.
Endoplasmic reticulum stress controls iron metabolism through TMPRSS6 repression and hepcidin mRNA stabilization by RNA-binding protein HuR.
Haematologica (2020-07-25)
Audrey Belot, Ophélie Gourbeyre, Anais Palin, Aude Rubio, Amélie Largounez, Céline Besson-Fournier, Chloé Latour, Megane Lorgouilloux, Inka Gallitz, Alexandra Montagner, Arnaud Polizzi, Marion Régnier, Sarra Smati, An-Sheng Zhang, Manuel D Diaz-Munoz, Andrea U Steinbicker, Hervé Guillou, Marie-Paule Roth, Hélène Coppin, Delphine Meynard
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
IgG, Rabbit, Rabbit IgG Polyclonal Antibody validated for use in ELISA.