Merck
Design and synthesis of novel antibacterial peptide-resin conjugates.
Bioorganic & medicinal chemistry letters (2007-09-11)
Won-Mi Cho, Bishnu Prasad Joshi, Hyeongjin Cho, Keun-Hyeung Lee
ABSTRAKT
We synthesized a novel peptide-resin conjugate by immobilizing beta-sheet antibacterial peptide on PEG-PS resin. The peptide-resin conjugate, similar to cationic antimicrobial peptides, demonstrated unique properties such as potent antibacterial activity, no hemolytic activity, lipid membrane perturbation activity, and potent synergism with vancomycin. Specially, the peptide-resin conjugate showed a more increased lipid membrane perturbation activity in comparison to unbound beta-sheet antibacterial peptide.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Melittin from honey bee venom, ≥65% (HPLC)
Sigma-Aldrich
Melittin, ≥97% (HPLC), synthetic
Sigma-Aldrich
Melittin from honey bee venom, ≥85% (HPLC)