- Mutation and inhibition studies of mevalonate 5-diphosphate decarboxylase.
Mutation and inhibition studies of mevalonate 5-diphosphate decarboxylase.
Bioorganic & medicinal chemistry letters (2007-09-25)
Yongge Qiu, Jinbo Gao, Fei Guo, Yuqin Qiao, Ding Li
PMID17888661
ABSTRAKT
Mevalonate 5-diphosphate decarboxylase plays an important role in regulating cholesterol biosynthesis, which was studied through incubation with various synthetic substrate analogs and characterization of mutated enzymes. The results are potentially useful for further developing inhibitors that block the mevalonate pathway which is a drug target for treating cardiovascular disease and cancer.