Merck
  • Discovery of (Z)-2-phenyl-3-(1H-pyrrol-2-yl)acrylonitrile derivatives active against Haemonchus contortus and Ctenocephalides felis (cat flea).
Discovery of (Z)-2-phenyl-3-(1H-pyrrol-2-yl)acrylonitrile derivatives active against Haemonchus contortus and Ctenocephalides felis (cat flea).
Bioorganic & medicinal chemistry letters (2006-12-08)
Abdelselam Ali, Marianne Bliese, Jo-Anne M Rasmussen, Roger M Sargent, Simon Saubern, David G Sawutz, John S Wilkie, David A Winkler, Kevin N Winzenberg, Ruth C J Woodgate
ABSTRAKT
A series of 2-phenyl-3-(1H-pyrrol-2-yl)acrylonitrile derivatives were synthesized and evaluated for in vitro activity against the endoparasite Haemonchus contortus and the ectoparasite Ctenocephalides felis. Some compounds had significant in vitro activity against these parasites.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ANI-7, ≥98% (HPLC)
Supelco
Closantel, PESTANAL®, analytical standard