Merck
Transformation of mu-opioid receptor agonists into biologically potent mu-opioid receptor antagonists.
Bioorganic & medicinal chemistry (2006-12-05)
Tingyou Li, Yunden Jinsmaa, Masahiro Nedachi, Anna Miyazaki, Yuko Tsuda, Akihiro Ambo, Yusuke Sasaki, Sharon D Bryant, Ewa Marczak, Qiang Li, H Scott Swartzwelder, Lawrence H Lazarus, Yoshio Okada
ABSTRAKT
N-Allylation (-CH(2)-CHCH(2)) of [Dmt(1)]endomorphins yielded the following: (i) [N-allyl-Dmt(1)]endomorphin-2 (Dmt=2',6'-dimethyl-l-tyrosine) (12) and [N-allyl-Dmt(1)]endomorphin-1 (15) (K(i)mu=0.45 and 0.26nM, respectively) became mu-antagonists (pA(2)=8.59 and 8.18, respectively) with weak delta-antagonism (pA(2)=6.32 and 7.32, respectively); (ii) intracerebroventricularly administered 12 inhibited morphine-induced CNS-mediated antinociception in mice [AD(50) (0.148ng/mouse) was 16-fold more potent than naloxone], but not spinal antinociception, and (iii) 15 reversed the alcohol-elevated frequency in spontaneous inhibitory post-synaptic currents (IPSC) in hippocampal CA1 pyramidal cells in rat brain slices (P=0.0055). Similarly, N-allylation of the potent mu-opioidmimetic agonists, 1,6-bis-[H-Dmt-NH]-hexane and 3,6-bis-[Dmt-NH-propyl]-2(1H)-pyrazinone, converted them into mu-antagonists (pA(2)=7.23 and 7.17 for the N-allyl-derivatives 17 and 19, respectively), and exhibited weak delta-antagonism. Thus, N-allylation of Dmt containing opioid peptides or opioidmimetics continues to provide a facile means to convert selective mu-opioid agonists into potent mu-opioid antagonists.