Merck
Cyclosporin A inhibits the propagation of influenza virus by interfering with a late event in the virus life cycle.
Japanese journal of infectious diseases (2013-07-26)
Itsuki Hamamoto, Kazuhiro Harazaki, Naohiko Inase, Hiroshi Takaku, Masato Tashiro, Norio Yamamoto
ABSTRAKT
Influenza is a global public health problem that causes a serious respiratory disease. Influenza virus frequently undergoes amino acid substitutions, which result in the emergence of drug-resistant viruses. To control influenza viruses that are resistant to currently available drugs, it is essential to develop new antiviral drugs with a novel molecular target. Here, we report that cyclosporin A (CsA) inhibits the propagation of influenza virus in A549 cells by interfering with a late event in the virus life cycle. CsA did not affect adsorption, internalization, viral RNA replication, or synthesis of viral proteins in A549 cells, but inhibited the step(s) after viral protein synthesis, such as assembly or budding. In addition, siRNA-mediated knockdown of the expression of the major CsA targets, namely cyclophilin A (CypA), cyclophilin B (CypB), and P-glycoprotein (Pgp), did not inhibit influenza virus propagation. These results suggest that CsA inhibits virus propagation by mechanism(s) independent of the inhibition of the function of CypA, CypB, and Pgp. CsA may target an unknown molecule that works as a positive regulator in the propagation of influenza virus. Our findings would contribute to the development of a novel anti-influenza virus therapy and clarification of the regulatory mechanism of influenza virus multiplication.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®