Merck
  • Modification of eukaryotic initiation factor 5A from Plasmodium vivax by a truncated deoxyhypusine synthase from Plasmodium falciparum: An enzyme with dual enzymatic properties.
Modification of eukaryotic initiation factor 5A from Plasmodium vivax by a truncated deoxyhypusine synthase from Plasmodium falciparum: An enzyme with dual enzymatic properties.
Bioorganic & medicinal chemistry (2007-06-27)
Annette Kaiser, Ina Hammels, Andrea Gottwald, Marwa Nassar, Mai Saad Zaghloul, Basma Abdal Motaal, Joachim Hauber, Achim Hoerauf
ABSTRAKT
The increasing resistance of the malaria parasites enforces alternative directions in finding new drug targets. Present findings from the malaria parasite Plasmodium vivax, causing tertiary malaria, suggest eukaryotic initiation factor 5A (eIF-5A) to be a promising target for the treatment of malaria. Previously we presented the 162 amino acid sequence of eukaryotic initiation factor 5A (eIF-5A) from Plasmodium vivax. In the present study, we have expressed and purified the 20kDa protein performed by one-step Nickel chelate chromatography. In Western blot experiments eIF-5A from P. vivax crossreacts with a polyclonal anti-eIF-5A antiserum from the plant Nicotiana plumbaginifolia (Solanaceae). Transcription of eIF-5A can be observed in both different developmental stages of the parasite being prominent in trophozoites. We recently published the nucleic acid sequence from a genomic clone of P. falciparum strain NF54 encoding a putative deoxyhypusine synthase (DHS), an enzyme that catalyzes the post-translational modification of eIF-5A. After removal of 22 amino acids DHS was expressed as a Histidin fusion protein and purified by Nickel affinity chromatography. Truncated DHS from P. falciparum modifies eIF-5A from P. vivax. DHS from P. falciparum NF54 is a bi-functional protein with dual enzymatic specificities, that is, DHS activity and homospermidine synthase activity (HSS) (0.047 pkatal/mg protein) like in other eukaryotes. Inhibition of DHS from P. falciparum resulted in a K(i) of 0.1 microM for the inhibitor GC7 being 2000-fold less than the nonguanylated derivative 1,7-diaminoheptane. Dhs transcription occurs in both develomental stages suggesting its necessity in cell proliferation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,7-Diaminoheptane, 98%