Merck
The study of chiral recognition on ibuprofen enantiomers by a fluorescent probe based on β-cyclodextrin modified ZnS:Mn quantum dots.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2020-10-10)
Yanyan Zeng, Yueting Wang, Zhihui Liang, Zhe Jiao
ABSTRAKT
In this paper, a fluorescence method for chiral detection of ibuprofen and its enantiomer was developed. The L-cystenine-capped ZnS:Mn quantum dots were synthesized and functionalized with β-cyclodextrin (β-CD-QDs). The β-CD-QDs exhibited different quenching effect to the S-(+)-ibuprofen and the R-(-)-ibuprofen based on the advantage of the inclusion complex of cyclodextrin. It was found that the quenching of β-CD-QDs by S-(+)-ibuprofen was due to the formation of inclusion complex through both static quenching and photoinduced electron transfer, but only slight quenching with the R-(-)-ibuprofen. The stability constants derived from Hildebrand-Benesi method and absorption titration experiments were applied to determine the stability constants of the formed complexes, the double reciprocal plots suggest that a conclusion complex with a ratio of 1:1 was formed between β-CD-QDs and S-(+)-ibuprofen, but did not with the R-(-)-ibuprofen. The fluorescence intensity of the β-CD-QDs was linearly dependent on the concentration of the S-(+)-IBP in the range of 0-0.5 nmol/L with an limit of detection of 0.29 nmol/L.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
p-Toluenesulfonyl chloride, ReagentPlus®, ≥99%