Merck
A VTA GABAergic Neural Circuit Mediates Visually Evoked Innate Defensive Responses.
Neuron (2019-06-17)
Zheng Zhou, Xuemei Liu, Shanping Chen, Zhijian Zhang, Yuanming Liu, Quentin Montardy, Yongqiang Tang, Pengfei Wei, Nan Liu, Lei Li, Ru Song, Juan Lai, Xiaobin He, Chen Chen, Guoqiang Bi, Guoping Feng, Fuqiang Xu, Liping Wang
ABSTRAKT
Innate defensive responses are essential for animal survival and are conserved across species. The ventral tegmental area (VTA) plays important roles in learned appetitive and aversive behaviors, but whether it plays a role in mediating or modulating innate defensive responses is currently unknown. We report that VTAGABA+ neurons respond to a looming stimulus. Inhibition of VTAGABA+ neurons reduced looming-evoked defensive flight behavior, and photoactivation of these neurons resulted in defense-like flight behavior. Using viral tracing and electrophysiological recordings, we show that VTAGABA+ neurons receive direct excitatory inputs from the superior colliculus (SC). Furthermore, we show that glutamatergic SC-VTA projections synapse onto VTAGABA+ neurons that project to the central nucleus of the amygdala (CeA) and that the CeA is involved in mediating the defensive behavior. Our findings demonstrate that aerial threat-related visual information is relayed to VTAGABA+ neurons mediating innate behavioral responses, suggesting a more general role of the VTA.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®