Merck
Dysregulated NF-κB-Dependent ICOSL Expression in Human Dendritic Cell Vaccines Impairs T-cell Responses in Patients with Melanoma.
Cancer immunology research (2020-10-15)
Deena M Maurer, Juraj Adamik, Patricia M Santos, Jian Shi, Michael R Shurin, John M Kirkwood, Walter J Storkus, Lisa H Butterfield, Deena M Maurer, Juraj Adamik, Patricia M Santos, Jian Shi, Michael R Shurin, John M Kirkwood, Walter J Storkus, Lisa H Butterfield
ABSTRAKT
Therapeutic cancer vaccines targeting melanoma-associated antigens are commonly immunogenic but are rarely effective in promoting objective clinical responses. To identify critical molecules for activation of effective antitumor immunity, we have profiled autologous dendritic cell (DC) vaccines used to treat 35 patients with melanoma. We showed that checkpoint molecules induced by ex vivo maturation correlated with in vivo DC vaccine activity. Melanoma patient DCs had reduced expression of cell surface inducible T-cell costimulator ligand (ICOSL) and had defective intrinsic NF-κB signaling. Chromatin immunoprecipitation assays revealed NF-κB-dependent transcriptional regulation of ICOSL expression by DCs. Blockade of ICOSL on DCs reduced priming of antigen-specific CD8+ and CD4+ T cells from naïve donors in vitro Concentration of extracellular/soluble ICOSL released from vaccine DCs positively correlated with patient clinical outcomes, which we showed to be partially regulated by ADAM10/17 sheddase activity. These data point to the critical role of canonical NF-κB signaling, the regulation of matrix metalloproteinases, and DC-derived ICOSL in the specific priming of cognate T-cell responses in the cancer setting. This study supports the implementation of targeted strategies to augment these pathways for improved immunotherapeutic outcomes in patients with cancer.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Ionomycin calcium salt from Streptomyces conglobatus, powder, ≥98% (HPLC)
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein