Merck
MicroRNA miR-7 Regulates Secretion of Insulin-Like Peptides.
Endocrinology (2019-12-27)
Pamela Agbu, Justin J Cassidy, Jonathan Braverman, Alec Jacobson, Richard W Carthew
ABSTRAKT
The insulin/insulin-like growth factor (IGF) pathway is essential for linking nutritional status to growth and metabolism. MicroRNAs (miRNAs) are short RNAs that are players in the regulation of this process. The miRNA miR-7 shows highly conserved expression in insulin-producing cells across the animal kingdom. However, its conserved functions in regulation of insulin-like peptides (ILPs) remain unknown. Using Drosophila as a model, we demonstrate that miR-7 limits ILP availability by inhibiting its production and secretion. Increasing miR-7 alters body growth and metabolism in an ILP-dependent manner, elevating circulating sugars and total body triglycerides, while decreasing animal growth. These effects are not due to direct targeting of ILP mRNA, but instead arise through alternate targets that affect the function of ILP-producing cells. The Drosophila F-actin capping protein alpha (CPA) is a direct target of miR-7, and knockdown of CPA in insulin-producing cells phenocopies the effects of miR-7 on ILP secretion. This regulation of CPA is conserved in mammals, with the mouse ortholog Capza1 also targeted by miR-7 in β-islet cells. Taken together, these results support a role for miR-7 regulation of an actin capping protein in insulin regulation, and highlight a conserved mechanism of action for an evolutionarily ancient microRNA.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Roche
Anty-HA-Peroksydaza, wysokie powinowactwo, from rat IgG1