Merck
A sensitive fluorescent probe based on dithizone-capped ZnS quantum dots for quercetin determination in biological samples.
Luminescence : the journal of biological and chemical luminescence (2020-06-28)
Susan Sadeghi, Mohadeseh Hosseinpour-Zaryabi
ABSTRAKT
A simple turn on/off fluorescence approach based on dithizone-capped ZnS quantum dots (ZnS@DZ QDs) with the help of lead ions as a fluorescent probe for the quantitative determination of quercetin is reported. The interaction of lead ions with dithizone led to the formation of a rigid structure on the surface of ZnS@DZ QDs and turned on the fluorescence intensity of the QDs. After addition of quercetin to this probe and interaction with lead ions, the fluorescence emission turned off. Concerning the quenching fluorescence intensity of ZnS@DZ QDs/Pb2+ QDs probe induced by the target, under the optimum conditions, the probe enabled detection of quercetin in the concentration range from 0.54 μM to 21.7 μM with a correlation coefficient of 0.993 and detection limit of 0.25 μM. The present probe was applied successfully to the determine quercetin as a nutritional biomarker in human serum and 24-h urine samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Cysteine, technical grade