Merck
Inhibition of human UDP-glucuronosyltransferase enzyme by belinostat: Implications for drug-drug interactions.
Toxicology letters (2020-12-09)
Xiaoyu Wang, Zhe Wang, Zhen Wang, Xiuyuan Chen, Hang Yin, Lili Jiang, Jun Cao, Yong Liu
ABSTRAKT
Belinostat is a pan-histone deacetylase (HDAC) inhibitor which recently approved for the treatment of relapsed/refractory Peripheral T-cell lymphomas (PTCL). To assess drug-drug interactions (DDIs) potential of belinostat via inhibition of UDP-glucuronosyltransferases (UGTs), the effects of belinostat on UGTs activities were investigated using the non-selective probe substrate 4-methylumbelliferone (4-MU) and trifluoperazine (TFP) by UPLC-MS/MS. Belinostat exhibited a wide range of inhibition against UGTs activities, particularly a potent non-competitive inhibition against UGT1A3, and weak inhibition against UGT1A1, 1A7, 1A8, 2B4 and 2B7. Further, in vitro-in vivo extrapolation (IVIVE) approaches were used to predict the risk of DDI arising from inhibition of UGTs. Our data indicate that the intravenous infusion of belinostat at clinical available dose can contribute a significant increase to the AUC of co-administrated drugs primarily cleared by UGT1A3 or UGT1A1, which will result in potential DDIs. In contrast, oral administrated belinostat is unlikely to cause significant DDIs through inhibition of glucuronidation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tri(2-furyl)phosphine, 99%