Merck
Altering model cell membranes by means of localized magnetic heating.
Colloids and surfaces. B, Biointerfaces (2020-08-21)
Silvia Ruiz-Rincón, Alejandro González-Orive, Valeria Grazú, Raluca M Fratila, Jesús M de la Fuente, Pilar Cea
ABSTRAKT
Isolated iron oxide magnetic nanoparticles (MNPs), 12 nm in diameter, coated with oleic acid molecules as capping agents have been deposited by the Langmuir-Blodgett (LB) method onto a model cell membrane incorporating 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) and Cholesterol (Chol) in the 1:1 ratio, which was also fabricated by the LB technique. Atomic Force Microscopy (AFM) experiments showed that the application of an alternating magnetic field results in the embedding of the MNPs through the phospholipidic layer. These experimental results reveal that the heating of individual MNPs may induce a local increase in the fluidity of the film with a large control of the spatial and temporal specificity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,2-Hexadecanediol, technical grade, 90%