Merck
  • Reaction of rosmarinic acid with nitrite ions in acidic conditions: discovery of nitro- and dinitrorosmarinic acids as new anti-HIV-1 agents.
Reaction of rosmarinic acid with nitrite ions in acidic conditions: discovery of nitro- and dinitrorosmarinic acids as new anti-HIV-1 agents.
Journal of medicinal chemistry (2008-03-21)
Mélanie Dubois, Fabrice Bailly, Gladys Mbemba, Jean-François Mouscadet, Zeger Debyser, Myriam Witvrouw, Philippe Cotelle
ABSTRAKT
Rosmarinic acid was reacted with nitrite ions under acidic conditions to give 6'-nitro- and 6',6''-dinitrorosmarinic acids according to the reaction time. Both compounds were active as HIV-1 integrase inhibitors at the submicromolar level. They also inhibited the viral replication in MT-4 cells with modest and similar selectivity indexes. The nitration of rosmarinic acid strongly improves the anti-integrase inhibition and the antiviral activity without increasing the cellular toxicity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rosmarinic acid, 96%