Merck
Catecholamines suppress fatty acid re-esterification and increase oxidation in white adipocytes via STAT3.
Nature metabolism (2020-07-23)
Shannon M Reilly, Chao-Wei Hung, Maryam Ahmadian, Peng Zhao, Omer Keinan, Andrew V Gomez, Julia H DeLuca, Benyamin Dadpey, Donald Lu, Jessica Zaid, BreAnne Poirier, Xiaoling Peng, Ruth T Yu, Michael Downes, Christopher Liddle, Ronald M Evans, Anne N Murphy, Alan R Saltiel
ABSTRAKT
Catecholamines stimulate the mobilization of stored triglycerides in adipocytes to provide fatty acids (FAs) for other tissues. However, a large proportion is taken back up and either oxidized or re-esterified. What controls the disposition of these FAs in adipocytes remains unknown. Here, we report that catecholamines redirect FAs for oxidation through the phosphorylation of signal transducer and activator of transcription 3 (STAT3). Adipocyte STAT3 is phosphorylated upon activation of β-adrenergic receptors, and in turn suppresses FA re-esterification to promote FA oxidation. Adipocyte-specific Stat3 KO mice exhibit normal rates of lipolysis, but exhibit defective lipolysis-driven oxidative metabolism, resulting in reduced energy expenditure and increased adiposity when they are on a high-fat diet. This previously unappreciated, non-genomic role of STAT3 explains how sympathetic activation can increase both lipolysis and FA oxidation in adipocytes, revealing a new regulatory axis in metabolism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody