Merck
  • (3R)-3-amino-4-(2,4,5-trifluorophenyl)-N-{4-[6-(2-methoxyethoxy)benzothiazol-2-yl]tetrahydropyran-4-yl}butanamide as a potent dipeptidyl peptidase IV inhibitor for the treatment of type 2 diabetes.
(3R)-3-amino-4-(2,4,5-trifluorophenyl)-N-{4-[6-(2-methoxyethoxy)benzothiazol-2-yl]tetrahydropyran-4-yl}butanamide as a potent dipeptidyl peptidase IV inhibitor for the treatment of type 2 diabetes.
Bioorganic & medicinal chemistry letters (2008-09-30)
Aiko Nitta, Hideaki Fujii, Satoshi Sakami, Yutaka Nishimura, Tomofumi Ohyama, Mikiya Satoh, Junko Nakaki, Shiho Satoh, Chifumi Inada, Hideki Kozono, Hiroki Kumagai, Masahiro Shimamura, Tominaga Fukazawa, Hideki Kawai
ABSTRAKT
Novel series of 3-amino-N-(4-aryl-1,1-dioxothian-4-yl)butanamides and 3-amino-N-(4-aryltetrahydropyran-4-yl)butanamides were synthesized and evaluated as dipeptidyl peptidase IV (DPP-IV) inhibitors. Derivatives incorporating the 6-substituted benzothiazole group showed highly potent DPP-IV inhibitory activity. Oral administration of (3R)-3-amino-4-(2,4,5-trifluorophenyl)-N-{4-[6-(2-methoxyethoxy)benzothiazol-2-yl]tetrahydropyran-4-yl}butanamide (12u) reduced blood glucose excursion in an oral glucose tolerance test.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Testosterone, VETRANAL®, analytical standard
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard