- Anticancer fungal natural products: Mechanisms of action and biosynthesis.
Anticancer fungal natural products: Mechanisms of action and biosynthesis.
European journal of medicinal chemistry (2020-07-12)
Siwen Yuan, Jannu Vinay Gopal, Shuya Ren, Litong Chen, Lan Liu, Zhizeng Gao
PMID32652407
ABSTRAKT
Many fungal metabolites show promising anticancer properties both in vitro and in animal models, and some synthetic analogs of those metabolites have progressed into clinical trials. However, currently, there are still no fungi-derived agents approved as anticancer drugs. Two potential reasons could be envisioned: 1) lacking a clear understanding of their anticancer mechanism of action, 2) unable to supply enough materials to support the preclinical and clinic developments. In this review, we will summarize recent efforts on elucidating the anticancer mechanisms and biosynthetic pathways of several promising anticancer fungal natural products.