Merck
  • NPR-C gene polymorphism is associated with increased susceptibility to coronary artery disease in Chinese Han population: a multicenter study.
NPR-C gene polymorphism is associated with increased susceptibility to coronary artery disease in Chinese Han population: a multicenter study.
Oncotarget (2016-05-19)
Qin Hu, Qiji Liu, Shasha Wang, Xi Zhen, Zhimian Zhang, Ruijuan Lv, Guihua Jiang, Zhiyong Ma, Hong He, Daqing Li, Xiaoling Liu, Fei Gao, Jifu Li, Li Li, Mei Zhang, Xiaoping Ji, Yuguo Chen, Daowen Wang, Dejia Huang, Aiqun Ma, Wei Huang, Yuxia Zhao, Yaoqin Gong, Cheng Zhang, Yun Zhang
ABSTRAKT
To find a new locus that confers significant susceptibility to CAD in Chinese Han population, a genome-wide association study in 200 "extreme individuals" from a Shandong cohort and a pathway-based candidate gene study from a Shanghai cohort (293 CAD/293 controls) were simultaneously performed. Amongst them, 13 SNPs associated with CAD were selected to conduct validation and replication studies in additional 3363 CAD patients and 3148 controls. A novel locus rs700926 in natriuretic peptide receptor C (NPR-C) was identified in Shandong and Hubei cohorts. Then rs700926 and other nine tag SNPs were genotyped in four geographically different populations (Shandong, Shaanxi, Hubei and Sichuan cohorts), and 6 SNPs (rs700926, rs1833529, rs2270915, rs17541471, rs3792758 and rs696831) showed stronger association with CAD, regardless of single or combined analysis. We further genotyped rs2270915 and 10 additional tag SNPs in a central China cohort and identified rs12697273 and rs10066436 as the loci associated with CAD. All these positive associations remained significant after adjustment for traditional risk factors of CAD. NPR-C gene SNPs significantly contribute to CAD susceptibility in the Chinese Han population.