- Generation of a human CDX2 knock-in reporter iPSC line (MHHi007-A-1) to model human trophoblast differentiation.
Generation of a human CDX2 knock-in reporter iPSC line (MHHi007-A-1) to model human trophoblast differentiation.
Stem cell research (2018-06-01)
Svitlana V Malysheva, Stephanie Wunderlich, Alexandra Haase, Gudrun Göhring, Ulrich Martin, Sylvia Merkert
PMID29852468
ABSTRAKT
Caudal-type homeobox 2 (CDX2) transcription factor is an important marker for early trophoblast lineages and intestinal epithelium. Due to its nuclear expression the immunostaining and sorting of viable CDX2pos cells is not possible. In this paper we report the generation and describe key characteristics of a CDX2Venus knock-in reporter hiPSC-cell line (MHHi007-A-1) which can serve as an in vitro tool to study human trophoblast and intestinal differentiation.