Merck
TNF is a key cytokine mediating neutrophil cytotoxic activity in breast cancer patients.
NPJ breast cancer (2016-04-20)
Elizabeth Comen, Paulina Wojnarowicz, Venkatraman E Seshan, Riddhi Shah, Courtney Coker, Larry Norton, Robert Benezra
ABSTRAKT
We have previously shown a novel antimetastatic role for neutrophils in the premetastatic lung of mice in models of breast cancer. Here we expand on those findings in the context of human breast cancer. We assessed the cytotoxicity of neutrophils from 90 newly diagnosed breast cancer patients, 24 ductal carcinoma in situ patients, 56 metastatic breast cancer patients, and 64 women with no history of cancer. We report that neutrophils from metastatic and newly diagnosed breast cancer patients are significantly more cytotoxic than neutrophils from cancer-free individuals. We hypothesized that tumor-secreted factors 'prime' neutrophils to become cytotoxic. To identify these factors we assayed for cytokines in serum from 54 breast cancer patients and 35 cancer-free controls. Tumor necrosis factor (TNFα), MCP-1 (CCL2), and IL1RA significantly correlated with cytotoxicity and directly stimulated neutrophil cytotoxicity ex vivo. RNA-seq analyses found protein kinase C iota (PRKCI) to be over expressed in patient neutrophils relative to neutrophils from cancer-free individuals. PRKCI has been implicated in NADPH oxidase assembly, required for neutrophil-mediated cell cytotoxicity. Treatment of human neutrophils with TNF-induced PRKCI expression and cytotoxicity in samples that had low basal levels of PRKCI expression. To date, this work is the first to demonstrate the cytotoxic role of neutrophils in the peripheral blood of a large cohort of breast cancer patients, and that select cytokines appear to mediate the stimulation of neutrophil cytotoxicity. Further functional studies are necessary to identify clinically relevant means of stimulating neutrophil cytotoxicity as an effective barrier against disease progression and metastasis.