Merck
  • Selection for high-level telithromycin resistance in Staphylococcus aureus yields mutants resulting from an rplB-to-rplV gene conversion-like event.
Selection for high-level telithromycin resistance in Staphylococcus aureus yields mutants resulting from an rplB-to-rplV gene conversion-like event.
Antimicrobial agents and chemotherapy (2008-01-16)
Daniel R Gentry, David J Holmes
ABSTRAKT
While most Staphylococcus aureus telithromycin-resistant mutants isolated in this study possessed duplications within rplV (encoding ribosomal protein L22), four isolates possessed insertions within rplV that were identical to a portion of the gene rplB (encoding ribosomal protein L2). This novel type of mutation is the result of an apparent gene conversion-like event.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg