Merck
Absence of loss of heterozygosity of BRCA1 in a renal tumor from a BRCA1 germline mutation carrier.
Familial cancer (2012-10-23)
Shaheen Alanee, Sohela Shah, Rajmohan Murali, Rohini Rau-Murthy, Kasmintan A Schrader, Kenneth Offit
ABSTRAKT
BRCA1 functions as a tumor suppressor gene and germline and somatic mutations in this gene have been shown to be associated with many types of cancer. We report the first tumor study of renal cell carcinoma in a carrier of the deleterious BRCA1 mutation-c.68_69delAG.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-BRCA1 (Ab-1) Mouse mAb (MS110), liquid, clone MS110, Calbiochem®