- Antimalarial activity of a new stilbene glycoside from Parthenocissus tricuspidata in mice.
Antimalarial activity of a new stilbene glycoside from Parthenocissus tricuspidata in mice.
Antimicrobial agents and chemotherapy (2008-07-16)
Won-Hwan Park, Sung-Jae Lee, Hyung-In Moon
PMID18625780
ABSTRAKT
A novel stilbene glycoside [piceid-(1-->6)-beta-d-glucopyranoside; PBG] from Parthenocissus tricuspidata was tested in vivo against Plasmodium berghei. PBG exhibited significant blood schizontocidal activity in a 4-day early infection, a repository evaluation, and an established infection, with a significant mean survival time comparable to that obtained with the standard drug, chloroquine (5 mg x kg(-1) x day(-1)).