Merck
In vitro activities at pH 5.0 and pH 7.0 and in vivo efficacy of flucytosine against Aspergillus fumigatus.
Antimicrobial agents and chemotherapy (2008-09-17)
Paul E Verweij, Debbie T A Te Dorsthorst, Willem H P Janssen, Jacques F G M Meis, Johan W Mouton
ABSTRAKT
The antifungal agent flucytosine was found to be active in vitro against Aspergillus fumigatus isolates when the MIC was determined at pH 5.0 instead of pH 7.0. The in vitro MIC at pH 5.0 corresponded to the in vivo efficacy of flucytosine monotherapy in a murine model of invasive aspergillosis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5-Fluorocytosine, nucleoside analog