Merck
Ibuprofen inhibited migration of skeletal muscle cells in association with downregulation of p130cas and CrkII expressions.
Skeletal muscle (2019-08-30)
Chih-Hao Liao, Li-Ping Lin, Tung-Yang Yu, Chih-Chin Hsu, Jong-Hwei S Pang, Wen-Chung Tsai
ABSTRAKT
Nonsteroidal anti-inflammatory drugs (NSAIDs) are commonly used to treat sports-related muscle injuries. However, NSAIDs were recently shown to impede the muscle healing process after acute injury. Migration of skeletal muscle cells is a crucial step during the muscle healing process. The present study was performed to investigate the effect and molecular mechanisms of action of ibuprofen, a commonly used NSAID, on the migration of skeletal muscle cells. Skeletal muscle cells isolated from the gastrocnemius muscle of Sprague-Dawley rats were treated with ibuprofen. MTT assay (3-[4,5-dimethylthiazol-2-yl]-2,5-diphenyltetrazolium bromide) was used to evaluate cell viability, and cell apoptosis was evaluated by TUNEL assay, after ibuprofen treatment. Skeletal muscle cell migration and spreading were evaluated using the transwell filter migration assay and F-actin staining, respectively. The protein expression of p130cas and CrkII, which are cell migration facilitating genes, was determined by western blot analysis. The overexpression of p130cas of muscle cells was achieved by p130cas vector transfection. The results demonstrated that ibuprofen did not have a significant negative effect on cell viability and apoptosis. Ibuprofen inhibited the migration and spreading of skeletal muscle cells in a dose-dependent manner. Ibuprofen also dose-dependently decreased the protein expression of p130cas and CrkII. Furthermore, overexpression of p130cas resulted in the promotion of cell migration and spreading and counteracted ibuprofen-mediated inhibition. This study suggested that ibuprofen exerts a potentially adverse effect on the migration of skeletal muscle cells by downregulating protein expression of p130cas and CrkII. These results indicate a possible mechanism underlying the possible negative effect of NSAIDs on muscle regeneration.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
ApopTag Fluorescein In Situ Apoptosis Detection Kit, The ApopTag Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a Fluorescein reporter molecule.