Merck
Sphingomyelin synthesis in helminths: a minireview.
Folia parasitologica (1998-12-30)
I Bankov, A Timanova, J Barrett
ABSTRAKT
Sphingolipids are a diverse and ubiquitous group of lipids. They are widely distributed in parasites and a number of novel forms have been described. Sphingolipid synthesis has been investigated in the malarial parasite, cestodes, digeneans and nematodes. Although there are differences in detail, the synthetic pathways involved are similar to those found in mammals.