Merck
Draft Genome Sequence of Salinivibrio sp. Strain EAGSL, a Biotechnologically Relevant Halophilic Microorganism.
Microbiology resource announcements (2020-10-24)
Erin M Gaffney, Matteo Grattieri, Shelley D Minteer
ABSTRAKT
The halophilic bacterium Salinivibrio sp. strain EAGSL was isolated from the Great Salt Lake (Utah) for use in microbial electrochemical technologies experiencing fluctuating salt concentrations. Genome sequencing was performed with Ion Torrent technology, and the assembled genome reported here is 3,234,770 bp with a GC content of 49.41%.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
GenElute Bacterial Genomic DNA Kits, sufficient for 70 purifications