Merck
PKCα and PKCδ activation regulates transcriptional activity and degradation of progesterone receptor in human astrocytoma cells.
Endocrinology (2014-12-17)
Aliesha González-Arenas, Miguel Ángel Peña-Ortiz, Valeria Hansberg-Pastor, Brenda Marquina-Sánchez, Noemi Baranda-Ávila, Karen Nava-Castro, Alejandro Cabrera-Wrooman, Jesús González-Jorge, Ignacio Camacho-Arroyo
ABSTRAKT
Progesterone regulates cancer cell proliferation and invasion through its receptors (PR-A and PR-B), whose phosphorylation modifies their transcriptional activity and induce their degradation. We identified by in silico analysis a putative residue (Ser400) in PR that might be phosphorylated by protein kinase C (PKC), a family of enzymes involved in the proliferation and infiltration of astrocytomas, the most frequent and aggressive brain tumors. A grade III human astrocytoma-derived cell line was used to study the role of PKC in PR phosphorylation, transcriptional activity, and degradation. Treatment with PKC activator [tetradecanoyl phorbol acetate (TPA)] increased PR phosphorylation in Ser400 after 5 minutes, which in turn induced PR transcriptional activity and its subsequent degradation by the 26S proteasome 3-5 hours after treatment. Silencing or inhibition of PKCα and PKCδ blocked PR phosphorylation and degradation induced by TPA. Both PR isoforms were associated with PKCα and reached the maximum association after 5 minutes of TPA addition. These data correlated with immunnofluorescence assays in which nuclear colocalization of PKCα with PR increased after TPA treatment. We observed a 2-fold increase in cell proliferation after PKC activation with TPA that was reduced with the PR antagonist, RU486. The PR S400A mutant revealed that this residue is essential for PKC-mediated PR phosphorylation and degradation. Our results show a key participation of PKCα and PKCδ in PR regulation and function.