Merck
  • An Erythritol-Sweetened Beverage Induces Satiety and Suppresses Ghrelin Compared to Aspartame in Healthy Non-Obese Subjects: A Pilot Study.
An Erythritol-Sweetened Beverage Induces Satiety and Suppresses Ghrelin Compared to Aspartame in Healthy Non-Obese Subjects: A Pilot Study.
Cureus (2020-11-17)
Zachary A Sorrentino, Garrett Smith, Lindsey Palm, Kartik Motwani, John Butterfield, Christian Archer, Rebecca Henderson, Coy Heldermon, Shiva Gautam, Mark L Brantly
ABSTRAKT
Despite the reduced caloric content of artificially sweetened beverages (ASBs) relative to those sweetened with sucrose, consumption of ASBs fail to consistently decrease the risk of obesity and associated diseases. This failure may be due to the inability of ASBs to effectively reduce appetite and hence overall caloric intake. A variety of non-nutritive sweeteners (NNS), however, remain to be screened for effectiveness in promoting satiety and reducing calorie consumption. Erythritol is well-tolerated, low-calorie sugar alcohol widely used as a sugar substitute. It is unique among NNS due to its low sweetness index relative to glucose, meaning that it is typically served at much higher concentrations than other common NNS. Animal and human studies have noted correlations between osmolarity, satiety, and levels of satiety hormones, independent of the effects of sweetness or nutritive value. We hypothesized that consumption of a beverage sweetened with erythritol to the sweetness and osmolarity of a common soft drink will improve self-reported satiety and more strongly affect the magnitude of changes in the hormone ghrelin than would an iso sweet beverage sweetened only with aspartame, a sweetener with a high sweetness index relative to glucose. Using a randomized double-blind crossover trial, we found that serum ghrelin was significantly decreased after consumption of an erythritol-sweetened beverage compared to aspartame. Likewise, consumption of the erythritol-sweetened beverage increased various measures of satiety in volunteers. Knowledge gained from this project demonstrates that high-osmolarity NNS may be useful in formulating ASBs that are satiating and low in calories.