Merck
Antioxidant and antiradical activities of resorcinarene tetranitroxides.
Bioorganic & medicinal chemistry letters (2009-02-12)
Andriy I Vovk, Alexander M Shivanyuk, Roman V Bugas, Oxana V Muzychka, Andriy K Melnyk
ABSTRAKT
Resorcinarene oxazines bearing four TEMPO fragments at the wide rim of the macrocycle were prepared through the aminomethylation of resorcinarene octols with 4-amino-TEMPO and formaldehyde. Tetra-TEMPO resorcinarenes are efficient scavengers of 1,1-diphenyl-2-picrylhydrazyl radicals. The model studies revealed that macrocyclic structure and intramolecular hydrogen bonding make considerable contribution to antiradical activity of these compounds. Tetra-TEMPO resorcinarenes show also superoxide dismutase-like activity and efficiently inhibit ABAP-induced peroxidation of linoleic acid.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%