Merck
The T cell CD6 receptor operates a multitask signalosome with opposite functions in T cell activation.
The Journal of experimental medicine (2020-10-31)
Daiki Mori, Claude Grégoire, Guillaume Voisinne, Javier Celis-Gutierrez, Rudy Aussel, Laura Girard, Mylène Camus, Marlène Marcellin, Jérémy Argenty, Odile Burlet-Schiltz, Frédéric Fiore, Anne Gonzalez de Peredo, Marie Malissen, Romain Roncagalli, Bernard Malissen
ABSTRAKT
To determine the respective contribution of the LAT transmembrane adaptor and CD5 and CD6 transmembrane receptors to early TCR signal propagation, diversification, and termination, we describe a CRISPR/Cas9-based platform that uses primary mouse T cells and permits establishment of the composition of their LAT, CD5, and CD6 signalosomes in only 4 mo using quantitative mass spectrometry. We confirmed that positive and negative functions can be solely assigned to the LAT and CD5 signalosomes, respectively. In contrast, the TCR-inducible CD6 signalosome comprised both positive (SLP-76, ZAP70, VAV1) and negative (UBASH3A/STS-2) regulators of T cell activation. Moreover, CD6 associated independently of TCR engagement to proteins that support its implication in inflammatory pathologies necessitating T cell transendothelial migration. The multifaceted role of CD6 unveiled here accounts for past difficulties in classifying it as a coinhibitor or costimulator. Congruent with our identification of UBASH3A within the CD6 signalosome and the view that CD6 constitutes a promising target for autoimmune disease treatment, single-nucleotide polymorphisms associated with human autoimmune diseases have been found in the Cd6 and Ubash3a genes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-CD5 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Przeciwciało przeciw fosfotyrozynie, klon ®, clone 4G10®, Upstate®, from mouse