Merck
  • Protein tyrosine phosphatase 1B inhibitory effects of depsidone and pseudodepsidone metabolites from the Antarctic lichen Stereocaulon alpinum.
Protein tyrosine phosphatase 1B inhibitory effects of depsidone and pseudodepsidone metabolites from the Antarctic lichen Stereocaulon alpinum.
Bioorganic & medicinal chemistry letters (2009-04-14)
Changon Seo, Jae Hak Sohn, Jong Seog Ahn, Joung Han Yim, Hong Kum Lee, Hyuncheol Oh
ABSTRAKT
Seven phenolic lichen metabolites (1-7) have been isolated from a methanol extract of the Antarctic lichen Stereocaulon alpinum by various chromatographic methods. The structures of these compounds were determined mainly by analysis of NMR spectroscopic data. A depsidone-type compound, lobaric acid (1) and two pseudodepsidone-type compounds, 2 and 3, exhibited potent inhibitory activity against protein tyrosine phosphatase 1B (PTP1B) with IC(50) values of 0.87microM, 6.86microM, and 2.48microM, respectively. Kinetic analyses of PTP1B inhibition by compounds 1 and 2 suggested that these compounds inhibited PTP1B activity in a non-competitive manner.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ursolic acid, ≥90%
Supelco
Ursolic acid, analytical standard