Merck
  • A microwave-assisted facile regioselective Fischer indole synthesis and antitubercular evaluation of novel 2-aryl-3,4-dihydro-2H-thieno[3,2-b]indoles.
A microwave-assisted facile regioselective Fischer indole synthesis and antitubercular evaluation of novel 2-aryl-3,4-dihydro-2H-thieno[3,2-b]indoles.
Bioorganic & medicinal chemistry letters (2009-05-01)
Subramanian Vedhanarayanan Karthikeyan, Subbu Perumal, Krithika Arun Shetty, Perumal Yogeeswari, Dharmarajan Sriram
ABSTRAKT
A series of novel 2-aryl-3,4-dihydro-2H-thieno[3,2-b]indoles has been synthesised regioselectively in good yields from the reaction of 5-aryldihydro-3(2H)-thiophenones and arylhydrazine hydrochloride. This reaction is found to be assisted by microwaves. The thieno[3,2-b]indoles were evaluated for their in vitro activity against Mycobacterium tuberculosis H37Rv (MTB) and multi-drug resistant M. tuberculosis (MDR-TB). Among 22 compounds screened, [2-(2,4-dichlorophenyl)-7-fluoro-3,4-dihydro-2H-thieno[3,2-b]indole] (6t) was found to the most active compound with MIC of 0.4 microg/mL against MTB and MDR-TB.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Pyrazinecarboxamide