- Gene-corrected human Munc13-4-deficient CD8+ T cells can efficiently restrict EBV-driven lymphoproliferation in immunodeficient mice.
Gene-corrected human Munc13-4-deficient CD8+ T cells can efficiently restrict EBV-driven lymphoproliferation in immunodeficient mice.
Blood (2016-11-02)
Tayebeh Soheili, Julie Rivière, Ida Ricciardelli, Amandine Durand, Els Verhoeyen, Anne-Céline Derrien, Chantal Lagresle-Peyrou, Geneviève de Saint Basile, François-Loïc Cosset, Persis Amrolia, Isabelle André-Schmutz, Marina Cavazzana
PMID27799161
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse