Merck
Regulation of T cell receptor beta gene rearrangements and allelic exclusion by the helix-loop-helix protein, E47.
Immunity (2007-12-21)
Yasutoshi Agata, Nobuyuki Tamaki, Shuji Sakamoto, Tomokatsu Ikawa, Kyoko Masuda, Hiroshi Kawamoto, Cornelis Murre
ABSTRAKT
Allelic exclusion of antigen-receptor genes is ensured primarily by monoallelic locus activation upon rearrangement and subsequently by feedback inhibition of continued rearrangement. Here, we demonstrated that the basic helix-loop-helix protein, E47, promoted T cell receptor beta (TCRbeta) gene rearrangement by directly binding to target gene segments to increase chromatin accessibility in a dosage-sensitive manner. Feedback signaling abrogated E47 binding, leading to a decline in accessibility. Conversely, enforced expression of E47 induced TCRbeta gene rearrangement by antagonizing feedback inhibition. Thus, the abundance of E47 is rate limiting in locus activation, and feedback signaling downregulates E47 activity to ensure allelic exclusion.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®