Merck
Generation of iPSC-lines from two independent Takotsubo syndrome patients with recurrent Takotsubo events.
Stem cell research (2020-04-18)
Daniela Hübscher, Sabine Rebs, Wiebke Maurer, Jelena R Ghadri, Ralf Dressel, Christian Templin, Katrin Streckfuss-Bömeke
ABSTRAKT
The Takotsubo syndrome (TTS) is characterized by acute transient left ventricular dysfunction in the absence of obstructive coronary lesions. An enhanced β-adrenergic signaling and higher sensitivity to catecholamine-induced-toxicity were identified as mechanisms associated with TTS. It is still elusive, whether TTS patients with recurrent events show similar underlying signaling pathomechanism. Induced pluripotent stem cell (iPSC)-lines were generated from skin fibroblasts of two independent female Takotsubo syndrome patients with a severe phenotype characterized by recurrent TTS events. For reprogramming, a non-integrative plasmid technique was used. All generated iPSCs maintained full pluripotency, genomic integrity, and spontaneous in vitro and in vivo differentiation capacity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid