Merck
The first Hes1 dimer inhibitors from natural products.
Bioorganic & medicinal chemistry letters (2009-09-01)
Midori A Arai, Ayako Masada, Toshiyuki Ohtsuka, Ryoichiro Kageyama, Masami Ishibashi
ABSTRAKT
In the present study, we developed a high-throughput screening system for small molecule-inhibitors of the basic helix-loop-helix (bHLH) transcriptional repressor factor Hes1. Successful dimerization of Hes1 immobilized on a microplate and fluorophore (Cy3)-labelled Hes1 was confirmed. Using this system, several natural products were identified as the first Hes1 dimer inhibitors. Of these, two compounds which were isolated from myxomycetes (true slime molds) inhibited Hes1 from N box-dependent suppression of the gene expression in C3H10T1/2 cells.