Merck
Effect of Multi-Phosphonate Coating of Titanium Surfaces on Osteogenic Potential.
Materials (Basel, Switzerland) (2020-12-23)
Christian Wehner, Christian Behm, Selma Husejnagic, Andreas Moritz, Xiaohui Rausch-Fan, Oleh Andrukhov
ABSTRAKT
The aim of this study was to evaluate the impact of a novel multi-phosphonate (MP) coating strategy of dental implant surfaces on the expression of osteogenesis-related factors in vitro. MG-63 human osteoblast-like cells, bone marrow mesenchymal stem cells (BM-MSCs), and human periodontal ligament stem cells (hPDLSCs) were cultured separately on titanium disks with and without MP coating. Cell attachment was visualized by focal adhesion and actin cytoskeleton staining. The proliferation and gene expression of the markers related to osteogenesis and bone turnover were measured after 48 and 120 h of cell culture. Actin cytoskeleton assembly and focal adhesion were similar between test surfaces within each cell type but differed from those on tissue culture plastic (TCP). The proliferation of MG-63 cells and PDLSCs was comparable on all surfaces, while BM-MSCs showed an increase on tissue culture plastic (TCP) versus titanium. The gene expression of osteoprotegerin and receptor activator of nuclear factor-kappa B ligand was higher in MG-63 cells grown on MP-coated surfaces. At the same time, osteocalcin was decreased compared to the other surfaces. Collagen type I gene expression after 120 h was significantly lower in hPDLSCs cultivated on MP-coated surfaces. Within the limitations of this study, MP coating on titanium surfaces might have a slight beneficial effect on bone turnover in vitro.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Actin Cytoskeleton / Focal Adhesion Staining Kit, The Actin Cytoskeleton & Focal Adhesion Staining Kit consists of TRITC-conjugated phalloidin, anti-Vinculin & DAPI for the immunofluorescent staining of actin filaments in the cytoskeleton as well as the nucleus of the cells.